Tisk…
Anti-LRRFIP1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Dodavatel: Bioss
BS-12439R-A555
BOSSBS-12439R-A555EA 25430 CZK
BOSSBS-12439R-A555
Anti-LRRFIP1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
LRRFIP1 is an 738 amino acid human protein whose rodent counterpart is known as Lrrfip1 (also designated FLAP in mouse). LRRFIP1 is also believed to control smooth muscle cell proliferation following arterial injury through PDGF-A repression. The N-terminus of LRRFIP1 shows high homology to the coiled-coil domain of FLAP, a protein which binds the leucine-rich repeat (LRR) of Flightless I, and the interaction of LRRFIP1 with the LRR of Flightless I has been confirmed. LRRFIP1 does not bind single-stranded DNA or RNA significantly and binds double-stranded DNA weakly. In contrast, LRRFIP1More Product Information