Tisk…

Anti-CK1g2 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-12425R
BOSSBS-12425REA 20650 CZK
BOSSBS-12425R
Anti-CK1g2 Rabbit Polyclonal Antibody
Protilátky
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. They can phosphorylate a large number of proteins. Participates in Wnt signaling.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CK1g2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR