Tisk…
Anti-IPMK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-12385R-FITC
BOSSBS-12385R-FITCEA 25630 CZK
BOSSBS-12385R-FITC
Anti-IPMK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Inositol polyphosphate multikinase (IPMK) belongs to the inositol phosphokinase (IPK) family and is characterized as having a broad substrate specificity. However, IPMK displays a preference for inositol-1,4,5-trisphosphate (Ins(1,4,5)P3) and inositol 1,3,4,6-tetrakisphosphate (Ins(1,3,4,6)P4). IPMK is ubiquitously expressed with the highest expression in skeletal muscle, liver, placenta, lung, peripheral blood leukocytes, kidney, spleen and colon. IPMK is localized to the nucleus, where it may play a role in the regulation of calcium release from intracellular stores and has been implicatedMore Product Information