Tisk…

Anti-TCP1A Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BS-12292R-CY7
BOSSBS-12292R-CY7EA 25630 CZK
BOSSBS-12292R-CY7
Anti-TCP1A Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Molecular chaperone; assists the folding of proteins upon ATP hydrolysis. As part of the BBS/CCT complex may play a role in the assembly of BBSome, a complex involved in ciliogenesis regulating transports vesicles to the cilia. Known to play a role, in vitro, in the folding of actin and tubulin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TCP1A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR