Tisk…
Anti-ZNF843 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Dodavatel: Bioss
BS-12225R-A488
BOSSBS-12225R-A488EA 25430 CZK
BOSSBS-12225R-A488
Anti-ZNF843 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
ZNF843 is a 348 amino acid protein that is encoded by a gene which maps to chromosome 16. Chromosome 16 encodes over 900 genes in approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16 through the CREBBP gene which encodes a critical CREB binding protein. SignsMore Product Information
Objednat
Produktová specifikace
Symbol antigenu ZNF843
Environmentally Preferable
Název antigenu Zinc Finger Protein 843
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 488
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 283933
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C