Tisk…
Anti-ZNF350 Rabbit Polyclonal Antibody
Dodavatel: Bioss
BS-12213R
BOSSBS-12213REA 20650 CZK
BOSSBS-12213R
Anti-ZNF350 Rabbit Polyclonal Antibody
Protilátky
The transcpritional repressor ZBRK1, also designated zinc finger and BRCA1-interacting protein with a KRAB domain 1 or zinc finger protein 350, belongs to the Krüppel C2H2-type zinc-finger protein family of proteins. ZBRK1 localizes mainly to the nucleus and may be associated with the nuclear matrix. It is a widely expressed protein that binds to BRCA1. ZBRK1 plays an important role in transcriptional regulation. Likely targets of gene regulation are DNA damage response genes, which effect the survival and growth control of cells. ZBRK1 contains an N-terminal KRAB domain, a C-terminal BRCA1-More Product Information