Tisk…
Anti-PDGFAA + PDGFBB Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Dodavatel: Bioss
BS-12157R-A680
BOSSBS-12157R-A680EA 25340 CZK
BOSSBS-12157R-A680
Anti-PDGFAA + PDGFBB Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterised by a motif of eight cysteines. This gene product can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located,More Product Information