Tisk…
Anti-PSGR Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Dodavatel: Bioss
BS-12076R-A647
BOSSBS-12076R-A647EA 25430 CZK
BOSSBS-12076R-A647
Anti-PSGR Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactoryMore Product Information