Tisk…

Anti-LPAR4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Dodavatel: Bioss
BS-12074R-A488
BOSSBS-12074R-A488EA 25430 CZK
BOSSBS-12074R-A488
Anti-LPAR4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
Receptor for lysophosphatidic acid (LPA), a mediator of diverse cellular activities. Transduces a signal by increasing the intracellular calcium ions and by stimulating adenylyl cyclase activity. The rank order of potency for agonists of this receptor is 1-oleoyl- >1-stearoyl- >1-palmitoyl- >1-myristoyl- >1-alkyl- >1-alkenyl-LPA (By similarity).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: LPAR4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu LPAR4
Environmentally Preferable
Název antigenu Lysophosphatidic acid receptor 4
Synonyma antigenu P2Y5-LIKE|P2RY9|GPR23|LPA4|P2Y9
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 488
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 78134
Izotyp IgG
Imunochemie Yes
Průtoková cytometrie Yes
Imunogen 177-203/370
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR