Tisk…

Anti-RAMP2 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-11971R
BOSSBS-11971REA 20050 CZK
BOSSBS-11971R
Anti-RAMP2 Rabbit Polyclonal Antibody
Protilátky
Transports the calcitonin gene-related peptide type 1 receptor (CALCRL) to the plasma membrane. Acts as a receptor for adrenomedullin (AM) together with CALCRL.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: RAMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu RAMP2
Název antigenu Receptor (G Protein-coupled) activity modifying protein 2
Environmentally Preferable
Synonyma antigenu RAMP2|Receptor activity-modifying protein 2|Calcitonin-receptor-like receptor activity-modifying protein 2|CRLR activity-modifying protein 2
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 10266
Izotyp IgG
Western blot Yes
Imunochemie Yes
Imunogen 90-145/175
Formulář liquid
Čištění Purified by Protein A.
Koncentrace 1 ug/ul
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Transportní teplota 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR