Tisk…
Anti-SLC5A3/SMIT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Dodavatel: Bioss
BS-11954R-A680
BOSSBS-11954R-A680EA 25340 CZK
BOSSBS-11954R-A680
Anti-SLC5A3/SMIT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
Myo-inositol is involved in many important aspects of cellular regulation including membrane structure, signal transduction and osmoregulation. It is taken up into cells by the sodium/myo-inositol cotransporter (SMIT). SMIT activity maintains intracellular concentrations of myo-inositol; it is upregulated in response to hypertonic stress. The human SMIT protein is encoded by the SLC5A3 gene, which maps to chromosome 21q22.12. It is expressed in many human tissues, such as brain, kidney and placenta. Specifically, SMIT is abundantly expressed throughout the whole brain and spinal cord inMore Product Information