Tisk…
Anti-NOR1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Dodavatel: Bioss
BS-11903R-A750
BOSSBS-11903R-A750EA 25340 CZK
BOSSBS-11903R-A750
Anti-NOR1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
Nur77 (also designated NGFI-B), Nurr1 (Nur-related factor 1), and NOR-1 (neuron-derived orphan receptor 1) constitute the NGFI-B subfamily within the nuclear receptor superfamily. Ligands for these protein have not been identified, and, therefore, they are designated orphan nuclear receptors. Genes of the NGFI-B subfamily are classified as immediate-early genes, which are induced rapidly, but transiently, in response to a variety of stimuli. They have been implicated in cell proliferation, differentiation, and apoptosis. The human NOR-1 gene maps to chromosome 9q, and encodes a protein whichMore Product Information