Tisk…
Anti-GHRHR Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-11794R-A350
BOSSBS-11794R-A350EA 38440 CZK
BOSSBS-11794R-A350
Anti-GHRHR Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
GHRH-R is a seven transmembrane domain protein that localizes to the somatotroph of the pituitary. GHRH-R plays an important role in growth and acts as a high-affinity receptor for GHRH. Binding of GHRH leads to the coupling of GHRH-R to G-protein which stimulates increased adenylyl cyclase activity and the accumulation of cAMP leading to the synthesis and release of growth hormone and somatotroph proliferation. In addition, this signalling pathway may have direct action in fetal/placental development, reproduction and immune function. GHRH and GHRH-R may also play a role in the regulationMore Product Information