Tisk…
Anti-REEP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-11752R-FITC
BOSSBS-11752R-FITCEA 25630 CZK
BOSSBS-11752R-FITC
Anti-REEP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Transport of G protein-coupled receptors (GPCRs) to the cell surface membrane is critical for receptor-ligand recognition. Mammalian GPCR odorant receptors (ORs), when heterologously expressed in cells, are poorly expressed on the cell surface. REEP1 (receptor expression-enhancing protein 1), is a 201 amino acid multi-pass mitochondrion membrane protein that belongs to the DP1 family. REEP1 interacts with odorant receptor proteins and may enhance the cell surface expression of odorant receptors. Mutations in the REEP1 gene are the third most common cause of hereditary spastic paraplegia (HSP)More Product Information