Tisk…
Anti-OR10J3 Rabbit Polyclonal Antibody (Cy7®)
Dodavatel: Bioss
BS-11629R-CY7
BOSSBS-11629R-CY7EA 25630 CZK
BOSSBS-11629R-CY7
Anti-OR10J3 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Olfactory receptors are G protein-coupled receptors that localize to the cilia of olfactory sensory neurons where they display affinity for and bind to a variety of odor molecules. The genes encoding olfactory receptors comprise the largest family in the human genome. The binding of olfactory receptor proteins to odor molecules triggers a signal transduction that propagates nerve impulses throughout the body, ultimately leading to transmission of the signal to the brain and the subsequent perception of smell. OR10J3 (olfactory receptor 10J3) is a 329 amino acid protein. The gene encoding OR10More Product Information
Objednat
Produktová specifikace
Symbol antigenu OR10J3
Název antigenu Olfactory Receptor, Family 10, Subfamily J Member 3
Environmentally Preferable
Synonyma antigenu O10J3_HUMAN|Olfactory receptor OR1 25 pseudogene|OR10J3|OR1 25|Olfactory receptor 10J3|Olfactory receptor family 10 subfamily J member 3 pseudogene|OR10J3P.|Olfactory receptor family 10 subfamily J member 3
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy7®
Reaktivita Human,Rat
Hostitel Rabbit
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C