Tisk…

Anti-OR5P3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BS-11628R-FITC
BOSSBS-11628R-FITCEA 25630 CZK
BOSSBS-11628R-FITC
Anti-OR5P3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Odorant receptor (Potential). May be involved in taste perception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OR5P3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR