Tisk…
Anti-OR2A4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-11624R-A350
BOSSBS-11624R-A350EA 25430 CZK
BOSSBS-11624R-A350
Anti-OR2A4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Olfactory receptors interact with odorant molecules in the nose to initiate a neuronal response that leads to the perception of smell. While they share a seven transmembrane domain structure with many neurotransmitter and hormone receptors, olfactory receptors are responsible for the recognition and transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. Both OR2A4 (olfactory receptor 2A4) and OR2A7 (olfactory receptor 2A7) are 310 amino acid multi-pass membrane proteins that belong to the G-protein coupled receptor 1 family. The gene that encodes OR2More Product Information
Objednat
Produktová specifikace
Symbol antigenu OR2A4
Environmentally Preferable
Název antigenu Olfactory Receptor, Family 2, Subfamily A Member 4
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 350
Reaktivita Human
Hostitel Rabbit
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C