Tisk…
Anti-NOMO1 Rabbit Polyclonal Antibody (Cy3®)
Dodavatel: Bioss
BS-11595R-CY3
BOSSBS-11595R-CY3EA 25630 CZK
BOSSBS-11595R-CY3
Anti-NOMO1 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Three highly similar proteins termed NOMO1, NOMO2 and NOMO3, are encoded by a gene mapping to a region of duplication on the p arm of human chromosome 16. All three NOMO proteins share similar functions and have been difficult to characterize individually. NOMO1 (Nodal modulator 1), also known as PM5, is a 1,222 amino acid highly conserved single-pass type I membrane protein expressed in colon tumor tissue and normal colonic mucosa. NOMO proteins are novel antagonists of Nodal signaling which interact with Nicalin to form a Nicalin-NOMO complex, and are rapidly degraded or stabilized byMore Product Information