Tisk…

Anti-HCRT Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BS-11520R-FITC
BOSSBS-11520R-FITCEA 25630 CZK
BOSSBS-11520R-FITC
Anti-HCRT Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Neuropeptides that play a significant role in the regulation of food intake and sleep-wakefulness, possibly by coordinating the complex behavioral and physiologic responses of these complementary homeostatic functions. A broader role in the homeostatic regulation of energy metabolism, autonomic function, hormonal balance and the regulation of body fluids, is also suggested. Orexin-A binds to both OX1R and OX2R with a high affinity, whereas orexin-B binds only to OX2R with a similar high affinity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HCRT
Clonality: Polyclonal
More Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR