Tisk…
Anti-CPT1C Rabbit Polyclonal Antibody (Cy5®)
Dodavatel: Bioss
BS-11416R-CY5
BOSSBS-11416R-CY5EA 25630 CZK
BOSSBS-11416R-CY5
Anti-CPT1C Rabbit Polyclonal Antibody (Cy5®)
Protilátky
The Cpt1 family of proteins are outer mitochondrial membrane proteins that regulate the entry into, and oxidation of fatty acids by, mitochondria. Malonyl-CoA, an intermediate in fatty acid synthesis, has been implicated as a regulatory component of the energy sensing system that feeds into hypothalmic neurons to impart energy homeostasis. Malonyl-CoA levels in the hypothalamus are dynamically regulated by fasting and feeding, altering subsequent feeding behaviour. Cpt1c, the brain-specific carnitine O-palmitoyltransferase 1, is thought to relay information about malonyl-CoA levels inMore Product Information