Tisk…
Anti-GPCR135/Relaxin 3 Receptor 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Dodavatel: Bioss
BS-11405R-A750
BOSSBS-11405R-A750EA 25340 CZK
BOSSBS-11405R-A750
Anti-GPCR135/Relaxin 3 Receptor 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
Relaxin Receptor 3 is a G protein-coupled receptor that binds Relaxin 3 and influences differentiation and maintenance of the nervous system. Relaxin Receptor 3 shares sequence similarity with somatostatin receptors and angiotensin receptors. It mediates central processing of sensory signals in the rat and is thought to be a modulator of stress responses. Relaxin Receptor 3 is present in the brain, with highest expression in substantia nigra and pituitary, followed by hippocampus, spinal cord, amygdala, caudate nucleus and corpus callosum, and low level expression in cerebellum. InMore Product Information