Tisk…

Anti-DLG1 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-11361R
BOSSBS-11361REA 20650 CZK
BOSSBS-11361R
Anti-DLG1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DLG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR