Tisk…
Anti-ICA1 Rabbit Polyclonal Antibody (Cy3®)
Dodavatel: Bioss
BS-11349R-CY3
BOSSBS-11349R-CY3EA 25630 CZK
BOSSBS-11349R-CY3
Anti-ICA1 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Carbonic anhydrases (CAs), also designated carbonate dehydratases or carbonate hydrolyases, form a large family of genes that encode zinc metalloenzymes of great physiologic importance. As catalysts of the reversible hydration of carbon dioxide, these enzymes participate in a variety of biologic processes, including respiration, acid-base balance, bone resorption and calcification as well as the formation of aqueous humor, cerebrospinal fluid, saliva and gastric acid. Genes in the ?carbonic anhydrase family encode either active carbonic anhydrase isozymes or 揳catalytic?(devoid of CO2More Product Information