Tisk…
Anti-PTPN5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-11328R-FITC
BOSSBS-11328R-FITCEA 25630 CZK
BOSSBS-11328R-FITC
Anti-PTPN5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
The brain-specific STEP (striatal enriched phosphatase) family of protein tyrosine phosphatases (PTPs) comprises both transmembrane and cytosolic protein members which are the products of alternative splicing. STEP family members are expressed in the dopaminoceptive neurons of the CNS, with highest expression in the basal ganglia and related structures. The STEP protein regulates the N-methyl-d-aspartate receptor (NMDAR) complex; STEP depresses both NMDAR single-channel activity and synaptic currents. The membrane-associated STEP61 isoform localizes in the postsynaptic densities (PSDs) ofMore Product Information
Objednat
Produktová specifikace
Symbol antigenu PTPN5
Název antigenu Protein Tyrosine Phosphatase, Non-receptor Type 5 (striatum-enriched)
Environmentally Preferable
Synonyma antigenu Striatum-enriched protein-tyrosine phosphatase|FLJ14427.|PTP STEP|Neural specic protein tyrosine phosphatase|Neural-specic protein-tyrosine phosphatase|STEP|PTPN 5|PTPSTEP|Tyrosine protein phosphatase non receptor type 5|Protein tyrosine phosphatase striatum enriched|Tyrosine-protein phosphatase non-receptor type 5|Ptpn5|PTN5_HUMAN|Protein tyrosine phosphatase non receptor type 5|Protein tyrosine phosphatase non receptor type 5 striatum enriched|PTN5
Druh protilátky Primární
Klonovalita Polyclonal
Spojení FITC (Fluorescein Isothiocyanate)
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C