Tisk…
Anti-PTPN5/STEP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Dodavatel: Bioss
BS-11328R-A750
BOSSBS-11328R-A750EA 25340 CZK
BOSSBS-11328R-A750
Anti-PTPN5/STEP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
The brain-specific STEP (striatal enriched phosphatase) family of protein tyrosine phosphatases (PTPs) comprises both transmembrane and cytosolic protein members which are the products of alternative splicing. STEP family members are expressed in the dopaminoceptive neurons of the CNS, with highest expression in the basal ganglia and related structures. The STEP protein regulates the N-methyl-d-aspartate receptor (NMDAR) complex; STEP depresses both NMDAR single-channel activity and synaptic currents. The membrane-associated STEP61 isoform localizes in the postsynaptic densities (PSDs) ofMore Product Information
Objednat
Produktová specifikace
Název antigenu PTPN5/STEP
Environmentally Preferable
Synonyma antigenu Striatum-enriched protein-tyrosine phosphatase|FLJ14427|PTP STEP|Neural specic protein tyrosine phosphatase|Neural-specic protein-tyrosine phosphatase|STEP|PTPN 5|PTPSTEP|Tyrosine protein phosphatase non receptor type 5|Protein tyrosine phosphatase striatum enriched|Tyrosine-protein phosphatase non-receptor type 5|Ptpn5|PTN5_HUMAN|Protein tyrosine phosphatase non receptor type 5|Protein tyrosine phosphatase non receptor type 5 striatum enriched|PTN5
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 750
Reaktivita Human,Rat
Hostitel Rabbit
Izotyp IgG
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Modifikace Unmodified
Epitop 201-300/565
Imunogen KLH conjugated synthetic peptide derived from human PTPN5
Formulář Liquid
Molekulární hmotnost 63 kDa
Čištění Purified by Protein A
Koncentrace 1 µg/µl
Skladovací pufr Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Skladovací teplota Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transportní teplota 4 °C
Testované aplikace ICC