Tisk…
Anti-ET-2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-11280R-A350
BOSSBS-11280R-A350EA 25430 CZK
BOSSBS-11280R-A350
Anti-ET-2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
The endothelin (ET) family of proteins, which includes ET-1 (endothelin-1), ET-2 (endothelin-2) and ET-3 (endothelin-3), are vasoactive peptides that are involved in various functions throughout the body. Endothelins can affect the central nervous system and neuronal excitability, and they elicit potent vasoconstrictor action. While ET-1 is a potent, 21-amino acid vasoconstrictor peptide, ET-2 has the most potent vasoconstrictor activity. ET-3 functions as a ligand for endothelin receptor type B (ETBR) and, through this interaction, mediates the maturation of enteric neurons and melanocytes.More Product Information