Tisk…

Anti-WFS1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Dodavatel: Bioss
BS-11272R-A680
BOSSBS-11272R-A680EA 25340 CZK
BOSSBS-11272R-A680
Anti-WFS1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
Participates in the regulation of cellular Ca²⁺ homeostasis, at least partly, by modulating the filling state of the endoplasmic reticulum Ca²⁺ store.

Type: Primary
Antigen: WFS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 791-890/890
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR