Tisk…

Anti-PAF Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BS-1119R-A350
BOSSBS-1119R-A350EA 25430 CZK
BOSSBS-1119R-A350
Anti-PAF Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Azelaoyl PAF is a potent PPARγ agonist which competes for the thiazolidinedione binding site.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Objednat

Produktová specifikace

Symbol antigenu PAF
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 350
Reaktivita Multiple
Hostitel Rabbit
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR