Tisk…
Anti-CASP12 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-1105R-FITC
BOSSBS-1105R-FITCEA 25890 CZK
BOSSBS-1105R-FITC
Anti-CASP12 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Caspases are cysteine proteases that cleave C-terminal aspartic acid residues on their substrate molecules. This gene is most highly related to members of the ICE subfamily of caspases that process inflammatory cytokines. In rodents, the homolog of this gene mediates apoptosis in response to endoplasmic reticulum stress. However, in humans this gene contains a polymorphism for the presence or absence of a premature stop codon. The majority of human individuals have the premature stop codon and produce a truncated non-functional protein. The read-through codon occurs primarily in individualsMore Product Information