Tisk…
Anti-Bestrophin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Dodavatel: Bioss
BS-11040R-A750
BOSSBS-11040R-A750EA 25340 CZK
BOSSBS-11040R-A750
Anti-Bestrophin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
Best vitelliform macular dystrophy, known as Best disease, is an early-onset autosomal dominant condition in which accumulation of lipofuscin-like material within and beneath the RPE leads to progressive loss of central vision. Best disease is frequently a reflection of mutations in the Bestrophin gene, which encodes a protein containing four putative transmembrane domains and localizes to the basolateral plasma membrane of RPE cells. Human Bestrophin forms oligomeric chloride channels that are sensitive to intracellular calcium. Missense mutations at the Bestrophin locus reduces orMore Product Information