Tisk…
Anti-KIF5A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dodavatel: Bioss
BS-11027R-HRP
BOSSBS-11027R-HRPEA 25630 CZK
BOSSBS-11027R-HRP
Anti-KIF5A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
NKHC1 is a neuronal-specific component of a multi-subunit “molecular motor” complex that mediates intracellular organelle transport. Mutations in the gene encoding NKHC1 cause autosomal dominant spastic paraplegia 10. NKHC1 has a pan-neuronal distribution in the nervous system. Rat tissue extracts by immunoblot of NKHC1 can produce a doublet only in brain and sciatic nerve tissue. NKHC1 is distributed throughout the central nervous system and is enriched in subsets of neurons. Within cultured hippocampal neurons, NKHC1 is concentrated in the perinuclear region of the cell body. KinesinMore Product Information