Tisk…

Anti-NPY1R Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BS-1070R-A555
BOSSBS-1070R-A555EA 25430 CZK
BOSSBS-1070R-A555
Anti-NPY1R Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is NPY >[Pro-34] PYY, PYY and [Leu-31, Pro-34] NPY >NPY (2-36) >[Ile-31, Gln-34] PP and PYY (3-36) >PP >NPY free acid.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPY1R
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR