Tisk…
Anti-PPP3R1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-10342R-A350
BOSSBS-10342R-A350EA 26230 CZK
BOSSBS-10342R-A350
Anti-PPP3R1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunit have been identified, designated PP1, PP2A, PP2B and PP2C. AnMore Product Information