Tisk…

Anti-ALDH1A1 Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BS-10162R-CY3
BOSSBS-10162R-CY3EA 25890 CZK
BOSSBS-10162R-CY3
Anti-ALDH1A1 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Binds free retinal and cellular retinol-binding protein-bound retinal. Can convert/oxidize retinaldehyde to retinoic acid (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALDH1A1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR