Tisk…

Anti-SCNN1D Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BS-10141R-CY5
BOSSBS-10141R-CY5EA 25630 CZK
BOSSBS-10141R-CY5
Anti-SCNN1D Rabbit Polyclonal Antibody (Cy5®)
Protilátky
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCNN1D
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu SCNN1D
Environmentally Preferable
Název antigenu Sodium Channel Nonvoltage-gated 1 Delta
Synonyma antigenu ENaCd|SCNED|ENaCdelta|dNaCh
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy5®
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 6339
Izotyp IgG
Imunochemie Yes
Imunogen 445-487/638
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR