Tisk…

Anti-SLC7A9 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BS-10085R-A350
BOSSBS-10085R-A350EA 38440 CZK
BOSSBS-10085R-A350
Anti-SLC7A9 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Involved in the high-affinity, sodium-independent transport of cystine and neutral and dibasic amino acids (system b(0,+)-like activity). Thought to be responsible for the high-affinity reabsorption of cystine in the kidney tubule.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC7A9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR