Tisk…

Anti-CD38 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BS-0980R-A647
BOSSBS-0980R-A647EA 25430 CZK
BOSSBS-0980R-A647
Anti-CD38 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Synthesizes the second messagers cyclic ADP-ribose and nicotinate-adenine dinucleotide phosphate, the former a second messenger for glucose-induced insulin secretion. Also has cADPr hydrolase activity. Also moonlights as a receptor in cells of the immune system.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR