Tisk…

Anti-CD38 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BS-0979R-HRP
BOSSBS-0979R-HRPEA 25630 CZK
BOSSBS-0979R-HRP
Anti-CD38 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Synthesizes the second messagers cyclic ADP-ribose and nicotinate-adenine dinucleotide phosphate, the former a second messenger for glucose-induced insulin secretion. Also has cADPr hydrolase activity (By similarity).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone: 0
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Produktová specifikace

Symbol antigenu CD38
Název antigenu CD38 molecule
Environmentally Preferable
Synonyma antigenu NAD+ nucleosidase|ADP-ribosyl cyclase 1|T10|cADPr hydrolase 1|ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase|cyclic ADP-ribose hydrolase 1|CD38 antigen p45
Druh protilátky Primární
Klonovalita Polyclonal
Spojení HRP (Horseradish Peroxidase)
Reaktivita Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 12494
Izotyp IgG
Western blot Yes
Imunochemie Yes
Imunogen 100-150/304
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR