Tisk…

Anti-PAG608 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BS-0942R-A350
BOSSBS-0942R-A350EA 25430 CZK
BOSSBS-0942R-A350
Anti-PAG608 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Acts as a bona fide target gene of p53/TP53. May play a role in the TP53-dependent growth regulatory pathway. May contribute to TP53-mediated apoptosis by regulation of TP53 expression and translocation to the nucleus and nucleolus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAG608
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR