Tisk…
Anti-ANGPT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-0800R-FITC
BOSSBS-0800R-FITCEA 25890 CZK
BOSSBS-0800R-FITC
Anti-ANGPT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme and inhibits endothelial permeability. The protein also contributes to blood vessel maturation and stability, and may be involved in earlyMore Product Information