Tisk…
Anti-ANGPT1 Rabbit Polyclonal Antibody (Cy5®)
Dodavatel: Bioss
BS-0800R-CY5
BOSSBS-0800R-CY5EA 25890 CZK
BOSSBS-0800R-CY5
Anti-ANGPT1 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme and inhibits endothelial permeability. The protein also contributes to blood vessel maturation and stability, and may be involved in earlyMore Product Information
Objednat
Produktová specifikace
Symbol antigenu ANGPT1
Environmentally Preferable
Název antigenu Angiopoietin 1
Synonyma antigenu Ang1|AGP1|Ang-1|1110046O21Rik
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy5®
Reaktivita Pig,Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 11600
Izotyp IgG
Imunochemie Yes
Imunogen 280-330/498
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C