Tisk…
Anti-SULT1E1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dodavatel: Bioss
BS-0629R-HRP
BOSSBS-0629R-HRPEA 25630 CZK
BOSSBS-0629R-HRP
Anti-SULT1E1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Sulfotransferase that utilizes 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor to catalyze the sulfate conjugation of estradiol and estrone. May play a role in the regulation of estrogen receptor activity by metabolizing free estradiol. Maximally sulfates beta-estradiol and estrone at concentrations of 2 nM. Also sulfates dehydroepiandrosterone, pregnenolone, ethinylestradiol, equalenin, diethylstilbesterol and 1-naphthol, at significantly higher concentrations; however, cortisol, testosterone and dopamine are not sulfated.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1More Product Information