Tisk…

Anti-ADIPOR1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Dodavatel: Bioss
BS-0610R-A488
BOSSBS-0610R-A488EA 25660 CZK
BOSSBS-0610R-A488
Anti-ADIPOR1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
Receptor for globular and full-length adiponectin (APM1), an essential hormone secreted by adipocytes that acts as an antidiabetic. Probably involved in metabolic pathways that regulate lipid metabolism such as fatty acid oxidation. Mediates increased AMPK, PPARA ligand activity, fatty acid oxidation and glucose uptake by adiponectin. Has some high-affinity receptor for globular adiponectin but low-affinity receptor for full-length adiponectin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADIPOR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host:More Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR