Tisk…

Anti-PTPRC Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BS-0522R-A555
BOSSBS-0522R-A555EA 25660 CZK
BOSSBS-0522R-A555
Anti-PTPRC Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
Protein tyrosine-protein phosphatase required for T-cell activation through the antigen receptor. Acts as a positive regulator of T-cell coactivation upon binding to DPP4. The first PTPase domain has enzymatic activity, while the second one seems to affect the substrate specificity of the first one. Upon T-cell activation, recruits and dephosphorylates SKAP1 and FYN. Dephosphorylates LYN, and thereby modulates LYN activity (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTPRC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR