Tisk…

Anti-Sodium / Hydrogen Exchanger 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodavatel: Bioss
BS-0505R-A750
BOSSBS-0505R-A750EA 25340 CZK
BOSSBS-0505R-A750
Anti-Sodium / Hydrogen Exchanger 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
Involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. Major proton extruding system driven by the inward sodium ion chemical gradient. Plays an important role in signal transduction.

Type: Primary
Antigen: Sodium / Hydrogen Exchanger 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 470-550/815
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR