Tisk…
Anti-Centromeric histone H3-like Protein-2 Rabbit Polyclonal Antibody
Dodavatel: Bioss
BS-0483R
BOSSBS-0483REA 20650 CZK
BOSSBS-0483R
Anti-Centromeric histone H3-like Protein-2 Rabbit Polyclonal Antibody
Protilátky
Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histone modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications can alter local chromatinMore Product Information