Tisk…
Anti-IgA Rabbit Polyclonal Antibody (Cy3®)
Dodavatel: Bioss
BS-0474R-CY3
BOSSBS-0474R-CY3EA 25630 CZK
BOSSBS-0474R-CY3
Anti-IgA Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Human IgA (immunoglobulin A) is a glycosylated protein of 160 kDa and is produced as a monomer or as a J chain linked dimer. Monomeric IgA constitutes 5-15 % of the serum immunoglobulins whereas dimeric IgA is localized to mucosa surfaces such as saliva, gastrointestinal secretion, bronchial fluids and milk. Mucosal IgA plays a major role in host defence by neutralising infectious agents at mucosal surfaces. The production is usually local and antigen specific IgA producing B cells can be found in regions under the lamina propria where they mature into dimeric IgA producing plasma cells. IgAMore Product Information
Objednat
Produktová specifikace
Symbol antigenu IgA
Environmentally Preferable
Název antigenu Immunoglobulin A
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy3®
Reaktivita Human
Hostitel Rabbit
Izotyp IgG
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C