Tisk…
Anti-MMP7 Rabbit Polyclonal Antibody (Cy3®)
Dodavatel: Bioss
BS-0423R-CY3
BOSSBS-0423R-CY3EA 25660 CZK
BOSSBS-0423R-CY3
Anti-MMP7 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Matrix metalloproteinase-7 (MMP-7) also known as matrilysin and PUMP (EC 3.4.24.23) cleaves a number of substrates including collagen types IV and X, elastin, fibronectin, gelatin, laminin and proteoglycans. MMP-7 is closely related to the stromelysin family members but is encoded by a different gene. MMP-7 is the smallest of all the MMPs consisting of a pro-peptide domain and a catalytic domain. It lacks the hemopexin-like domain common to other members of the MMPs. MMP-7 is secreted as a 28kD proenzyme and can be activated in vitro by organomercurials and trypsin and in vivo by MMP-3 to a 1More Product Information