Tisk…
Anti-GAD2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-0400R-A350
BOSSBS-0400R-A350EA 24950 CZK
BOSSBS-0400R-A350
Anti-GAD2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
This gene encodes one of several forms of glutamic acid decarboxylase, identified as a major autoantigen in insulin-dependent diabetes. The enzyme encoded is responsible for catalyzing the production of gamma-aminobutyric acid from L-glutamic acid. A pathogenic role for this enzyme has been identified in the human pancreas since it has been identified as an autoantibody and an autoreactive T cell target in insulin-dependent diabetes. This gene may also play a role in the stiff man syndrome. Alternative splicing results in multiple transcript variants that encode the same protein. [providedMore Product Information