Tisk…

Anti-F8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BS-0332R-A350
BOSSBS-0332R-A350EA 25660 CZK
BOSSBS-0332R-A350
Anti-F8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Factor VIII, along with calcium and phospholipid, acts as a cofactor for factor IXa when it converts factor X to the activated form, factor Xa.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: factor VIII/FVIII
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu F8
Název antigenu Coagulation Factor 8
Environmentally Preferable
Synonyma antigenu coagulation factor VIII|FVIII|DXS1253E|F8C|RP11-115M6.7|HEMA,antihemophilic factor|factor VIII F8B|AHF|coagulation factor VIIIc|F8B
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 350
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 2157
Izotyp IgG
Imunochemie Yes
ImunoFluorescence Yes
Imunogen 1470-1520/2351
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR